2-Bromo-N-cyclohexyl-5-fluorobenzamide

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

CA-4446-5G CA-4446-1G
COBBCA-4446-5GEA 0 GBP
COBBCA-4446-5G COBBCA-4446-1G
2-Bromo-N-cyclohexyl-5-fluorobenzamide
2-Bromo-N-cyclohexyl-5-fluorobenzamide
Formula: C₁₃H₁₅BrFNO
MDL Number: MFCD09800915
CAS Number: 951884-99-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR