4-Bromo-2-fluorobenzamide

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

OT-1003-5G OT-1003-1G
COBBOT-1003-5GEA 0 GBP
COBBOT-1003-5G COBBOT-1003-1G
4-Bromo-2-fluorobenzamide
4-Bromo-2-fluorobenzamide
Formula: C₇H₅BrFNO
MW: 218.03 g/mol
MDL Number: MFCD03094432
CAS Number: 292621-45-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR