3-Bromo-5-nitrobenzhydrazide
Supplier: COMBI-BLOCKS
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
CA-4573-5G
CA-4573-1G
COBBCA-4573-5GEA
0
GBP
COBBCA-4573-5G
COBBCA-4573-1G
3-Bromo-5-nitrobenzhydrazide
3-Bromo-5-nitrobenzhydrazide
Formula:
C₇H₆BrN₃O₃ |
MDL Number:
MFCD09972132 CAS Number: 374671-07-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...