4-Bromo-3-methoxybenzamide

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

CA-4370-1G CA-4370-5G
COBBCA-4370-1GEA 0 GBP
COBBCA-4370-1G COBBCA-4370-5G
4-Bromo-3-methoxybenzamide
4-Bromo-3-methoxybenzamide
Formula: C₈H₈BrNO₂
MDL Number: MFCD09475887
CAS Number: 176961-57-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR