Potassium-4-carboxyphenyltrifluoroborate

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: Potassium 4-carboxyphenyltrifluoroborate, Potassium (4-carboxyphenyl)trifluoroborate

TB-3202-5G TB-3202-25G TB-3202-1G
COBBTB-3202-5GEA 0 GBP
COBBTB-3202-5G COBBTB-3202-25G COBBTB-3202-1G
Potassium-4-carboxyphenyltrifluoroborate
Potassium 4-carboxyphenyltrifluoroborate
Formula: C₇H₅BF₃KO₂
MDL Number: MFCD04115737
CAS Number: 850623-38-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR