Potassium-3-(methoxycarbonyl)-5-nitrophenyltrifluoroborate

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

TB-3219-1G TB-3219-5G
COBBTB-3219-1GEA 0 GBP
COBBTB-3219-1G COBBTB-3219-5G
Potassium-3-(methoxycarbonyl)-5-nitrophenyltrifluoroborate
Potassium 3-(methoxycarbonyl)-5-nitrophenyltrifluoroborate
Formula: C₈H₆BF₃KNO₄
MDL Number: MFCD04115749
CAS Number: 850623-56-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR