Potassium-4-(methoxycarbonyl)phenyltrifluoroborate 96%
Supplier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
Synonyms:
Potassium trifluoro(4-(methoxycarbonyl)phenyl)borate
H52588.03
H52588.06
H52588.03EA
0
GBP
H52588.03
H52588.06
Potassium-4-(methoxycarbonyl)phenyltrifluoroborate 96%
Potassium 4-(methoxycarbonyl)phenyltrifluoroborate
Purity: 96%
CAS No.: 705254-34-8
Molecular Formula: C8H7BF3KO2
Formula Weight: 242.05
Melting Point:
Formula:
C₈H₇BF₃KO₂ Melting Pt: >300 °C Storage Temperature: Ambient |
MDL Number:
MFCD04115741 CAS Number: 705254-34-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...