3-Amino-3-(4-fluorophenyl)propionic acid

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

010001-1G 010001-5G
FLUO010001-1GEA 0 GBP
FLUO010001-1G FLUO010001-5G
3-Amino-3-(4-fluorophenyl)propionic acid
3-Amino-3-(4-fluorophenyl)propionic acid
Formula: C₉H₁₀FNO₂
MDL Number: MFCD00181811
CAS Number: 325-89-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR