3-Amino-3-(3-fluorophenyl)propionic acid

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 3-Amino-3-(3-fluorophenyl)propanoic acid

010740-1G 010740-10G 010740-5G
FLUO010740-1GEA 0 GBP
FLUO010740-1G FLUO010740-10G FLUO010740-5G
3-Amino-3-(3-fluorophenyl)propionic acid
3-Amino-3-(3-fluorophenyl)propionic acid
Formula: C₉H₁₀FNO₂
MDL Number: MFCD00764457
CAS Number: 117391-51-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR