4'-Iodo-2'-(trifluoromethyl)acetanilide

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

006673-1G 006673-5G
FLUO006673-1GEA 0 GBP
FLUO006673-1G FLUO006673-5G
4'-Iodo-2'-(trifluoromethyl)acetanilide
4'-Iodo-2'-(trifluoromethyl)acetanilide
Formula: C₉H₇F₃INO
MDL Number: MFCD00052845
CAS Number: 97760-98-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR