2,4-Dimethyl-5-thiazolecarboxylic acid

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

CFF387.5 CFF387.10 CFF387.25 017412-5G 017412-10G 017412-25G 017412-1G
CHMPCFF387.5EA 0 GBP
CHMPCFF387.5 CHMPCFF387.10 CHMPCFF387.25 FLUO017412-5G FLUO017412-10G FLUO017412-25G FLUO017412-1G
2,4-Dimethyl-5-thiazolecarboxylic acid
2,4-Dimethyl-5-thiazolecarboxylic acid
Formula: C₆H₇NO₂S
MDL Number: MFCD00052943
CAS Number: 53137-27-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR