Ethyl-2-amino-4-methylthiazole-5-carboxylate

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: Ethyl 2-amino-4-methylthiazole-5-carboxylate

011167-100G 011167-25G
FLUO011167-100GEA 0 GBP
FLUO011167-100G FLUO011167-25G
Ethyl-2-amino-4-methylthiazole-5-carboxylate
Ethyl 2-amino-4-methylthiazole-5-carboxylate
Formula: C₇H₁₀N₂O₂S
MDL Number: MFCD00123414
CAS Number: 7210-76-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR