2-Acetamidothiazole

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

013388-25G 013388-5G
FLUO013388-25GEA 0 GBP
FLUO013388-25G FLUO013388-5G
2-Acetamidothiazole
2-Acetamidothiazole
Formula: C₅H₆N₂OS
MW: 142.18 g/mol
MDL Number: MFCD00016892
CAS Number: 2719-23-5
EINECS: 220-321-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR