3-Benzoyl-2-thiophenecarboxylic acid
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
011433-500MG
011433-1G
FLUO011433-500MGEA
0
GBP
FLUO011433-500MG
FLUO011433-1G
3-Benzoyl-2-thiophenecarboxylic acid
3-Benzoyl-2-thiophenecarboxylic acid
Formula:
C₁₂H₈O₃S MW: 232.26 g/mol |
MDL Number:
MFCD00159542 CAS Number: 30006-03-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...