5-Bromo-4-(3-methoxyphenyl)pyrimidine
Supplier: COMBI-BLOCKS
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
HC-6123-5G
HC-6123-1G
COBBHC-6123-5GEA
0
GBP
COBBHC-6123-5G
COBBHC-6123-1G
5-Bromo-4-(3-methoxyphenyl)pyrimidine
5-Bromo-4-(3-methoxyphenyl)pyrimidine
Formula:
C₁₁H₉BrN₂O |
MDL Number:
MFCD09800945 CAS Number: 941294-40-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...