5-Bromo-4-phenylpyrimidine
Supplier: COMBI-BLOCKS
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
HC-6170-5G
HC-6170-1G
COBBHC-6170-5GEA
0
GBP
COBBHC-6170-5G
COBBHC-6170-1G
5-Bromo-4-phenylpyrimidine
5-Bromo-4-phenylpyrimidine
Formula:
C₁₀H₇BrN₂ MW: 235.0800 g/mol |
MDL Number:
MFCD00234599 CAS Number: 3543-46-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...