5-Bromo-4-(2,4-dimethylphenyl)pyrimidine
Supplier: COMBI-BLOCKS
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
HC-6122-1G
HC-6122-5G
COBBHC-6122-1GEA
0
GBP
COBBHC-6122-1G
COBBHC-6122-5G
5-Bromo-4-(2,4-dimethylphenyl)pyrimidine
5-Bromo-4-(2,4-dimethylphenyl)pyrimidine
Formula:
C₁₂H₁₁BrN₂ |
MDL Number:
MFCD09800944 CAS Number: 941294-39-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...