6-Bromo[1,2,4]triazolo[4,3-a]pyridine-3-thiol
Supplier: COMBI-BLOCKS
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
PY-7123-1G
PY-7123-250M
COBBPY-7123-1GEA
0
GBP
COBBPY-7123-1G
COBBPY-7123-250M
6-Bromo[1,2,4]triazolo[4,3-a]pyridine-3-thiol
6-Bromo-[1,2,4]triazolo[4,3-a]pyridine-3-thiol
Formula:
C₆H₄BrN₃S |
MDL Number:
MFCD11504977 CAS Number: 1093092-64-8 |
Learn more
![](/m-uk.vwr.com/en_GB/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...