6-Bromo[1,2,4]triazolo[1,5-a]pyridine

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 6-Bromo-[1,2,4]triazolo[1,5-a]pyridine

HI-1262-5G HI-1262-1G
COBBHI-1262-5GEA 0 GBP
COBBHI-1262-5G COBBHI-1262-1G
6-Bromo[1,2,4]triazolo[1,5-a]pyridine
6-Bromo-[1,2,4]triazolo[1,5-a]pyridine
Formula: C₆H₄BrN₃
MW: 198.02 g/mol
MDL Number: MFCD08689534
CAS Number: 356560-80-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR