2-Amino-4-(3-pyridyl)thiazole

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

CFF405.1 CFF405.2.5 CFF405.5 017428-1G 017428-2.5G 017428-5G
CHMPCFF405.1EA 0 GBP
CHMPCFF405.1 CHMPCFF405.2.5 CHMPCFF405.5 FLUO017428-1G FLUO017428-2.5G FLUO017428-5G
2-Amino-4-(3-pyridyl)thiazole
2-Amino-4-(3-pyridyl)thiazole
Formula: C₈H₇N₃S
MDL Number: MFCD00218013
CAS Number: 30235-27-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR