1-(2-Furoyl)piperazine

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: Prazosin Related Compound D

009787-25G 009787-100G
FLUO009787-25GEA 0 GBP
FLUO009787-25G FLUO009787-100G
1-(2-Furoyl)piperazine
1-(2-Furoyl)piperazine
Formula: C₉H₁₂N₂O₂
MDL Number: MFCD00038831
CAS Number: 40172-95-0
EINECS: 254-823-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR