Bis(triphenylphosphine)nickel(II) iodide
Supplier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Danger
|
Synonyms:
Diiodobis(triphenylphosphine)nickel(II)
18869.04
18869.09
18869.04EA
59.6
GBP
18869.04
18869.09
Bis(triphenylphosphine)nickel(II) iodide
Bis(triphenylphosphine)nickel (II) iodide
CAS No.: 82308-17-6
Molecular Formula: C36H30I2NiP2
Formula Weight: 837.08
Physical Form: Crystalline powder
Sensitivity: Air and Light Sensitive
Appearance: Dark brown
MDL No.: MFCD00015866
Formula:
C₃₆H₃₀I₂NiP₂ MW: 837.08 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00015866 CAS Number: 82308-17-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...