Bis(tetramethylcyclopentadienyl)nickel(II) ≥98%
Supplier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Danger
|
Synonyms:
Bis(tetramethylcyclopentadienyl)nickel
H27784.06
H27784.03
H27784.06EA
578
GBP
H27784.06
H27784.03
Bis(tetramethylcyclopentadienyl)nickel(II) ≥98%
Bis(tetramethylcyclopentadienyl)nickel (II)
Purity: 98+%
CAS No.: 79019-60-6
Molecular Formula: C18H26Ni
Formula Weight: 301.10
Storage Temperature: 2°C to 8°C
Melting Point: 107-112°
MDL No.: MFCD01862460
Formula:
C₁₈H₂₆Ni Melting Pt: 107…112 °C |
MDL Number:
MFCD01862460 CAS Number: 79019-60-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...