Tris(2,2,6,6-tetramethyl-3,5-heptanedionato-O,O')thulium(III) ≥98% (99.9%-Tm)
Supplier: Strem Chemicals, Inc.
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
69-7000.0001
69-7000.0005
STRE69-7000.0001EA
56.49
GBP
STRE69-7000.0001
STRE69-7000.0005
Tris(2,2,6,6-tetramethyl-3,5-heptanedionato-O,O')thulium(III) ≥98% (99.9%-Tm)
Tris(2,2,6,6-tetramethyl-3,5-heptanedionato-O,O')thulium(III)
Metal Beta-diketonates, Metal TMHD, Volatile Organometallics for CVD & ALD, Volatile Precursors for CVD
Formula:
C₃₃H₅₇O₆Tm Boiling Pt: 280 °C (dec.) Melting Pt: 169…172 °C |
MDL Number:
MFCD00015712 CAS Number: 15631-58-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...