CPC® Twist-to-Connect Quick-Disconnect Fitting, Sealing Cap, Polysulfone, MPU32039; 10/PK
Supplier: Avantor Fluid Handling
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
VWR Item#MFLX31310-02BLK
Your Organization's Price:
Retrieving
Twist-to-Connect locking mechanism protects against accidental disconnects.
  • Shrouded, leak-free seal protects against contamination
  • Provides rapid media transfer with smooth inner bore
These CPC® Twist-to-Connect quick-disconnect fittings feature an easy-to-use locking mechanism that guards against accidental disconnects and provides a reliable, secure connection. To assemble, place the insert in the coupling body and twist to lock. An audible click ensures your connection. A smooth inner bore and leak-free seal protect your process against contamination. All couplings are made from USP Class VI-compliant polysulfone with platinum-cured silicone seals. <br /><br />A complete coupler requires a fitting body and an insert (order separately).