Value Plastics® Fitting, Polypropylene, Straight, Female Luer x 1/8-27 NPT; 1000/PK
Supplier: Avantor Fluid Handling
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
VWR Item#MFLX30800-00BLK
Your Organization's Price:
Retrieving
Easily transition between luer fittings and UNF or NPT threads.
  • Determine which connector type is needed for your application by contacting free technical application assistance*
  • Global market leader in range of materials for most extensive chemical compatibility
  • Single-use custom configuration solutions to minimise downtime
  • Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
  • Connection Type: Female luer to 1/8-27 NPT thread
These adapters transition between luer fittings and UNF or NPT threads. Both male and female luer connections are available. All threaded connections are female thread.