Masterflex® Large Bore Female Luer ³⁄₁₆" Int.Ø in Natural Nylon Cleanroom Manufactured; 10 pack
Supplier: Avantor Fluid Handling
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
VWR Item#MFLX40310-04
Your Organization's Price:
Retrieving
These large-bore adapters are perfect for handling higher flow volumes in process while still using a slip-lock-style fitting system.