Masterflex® B/T® Analog Drive, Washdown, 12 to 321 rpm; 230 VAC
Supplier: Avantor Fluid Handling
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
VWR Item#MFLX77111-67-EU
Your Organization's Price:
Retrieving
High-volume transfer and dispensing.
  • Flow rate 0.71/2.1 to 42 LPM with B/T tubing—flow rate depends on drive rpm and tubing size
  • Rapid-Load® pump head occlusion mechanism slides occlusion bed out of the way for easy tube loading and improved tubing occlusion
  • Accepts B/T PerfectPosition™ pump tubing sizes B/T 87 and B/T 91
  • Interlock switch shuts off power to stop drive when door is opened
  • Detachable controller with 6-ft (1.9-m) cord mounts on pump or wall
  • IP56-rated motor protects against dust and strong jets of water
  • Reversible motor: purge before or after pumping, pump fluid in either direction
  • Includes 6-ft line cord with European plug
  • Power requirement for drive is 200 to 260 VAC, 50 Hz
  • Detachable controller with 6-ft (1.9-m) cord mounts on pump or wall
Masterflex® B/T® drives are the best choice for high-flow bulk-transfer applications. Their durable construction ensures long service life with minimal maintenance. The sturdy materials of construction include stainless steel/carbon steel, a cast aluminum housing with an epoxy-powder coating, and Nylatron® rollers for rugged dependability.<br /><br />All B/T drives include a new and improved Rapid-Load® pump head for fast and easy tubing changes. The redesigned occlusion mechanism provides an improved tubing retention mechanism which holds the tubing firmly in place to further simplify the loading process. PerfectPosition™ tubing sizes provide improved performance in higher-pressure applications and provide longer tubing life.
<b>Delivery information: </b>Includes rapid-Load pump head and 3 ft (1 m) of B/T 91 tubing. Order additional tubing under the Masterflex tubing section, B/T PerfectPosition Tubing.
<b>Delivery information: </b>Includes rapid-load pump head and 3 ft (1 m) of B/T 91 tubing.