LeviFlow LFS-10SU-Z Single-Use Precision Ultrasonic Flow Sensor, 20 L/min
Supplier: Avantor Fluid Handling
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
VWR Item#MFLX24500-14
Your Organization's Price:
Retrieving
Obtain flow measurements of high-purity fluids.
  • Order the starter kit and a flow sensor to build a complete system
  • Choose from sterile or non-sterile flow sensors covering a range of 80 ml/min to 80 LPM
  • Avoid contamination— systems don't contain moving or obtrusive parts
  • Sensors feature integrated sanitary clamp fittings
  • Easily configure flow sensor parameters via software
These ultrasonic flowmeter systems are designed for use in applications were cleanliness is required. Readings are accurate to 1%—high precision for high-purity fluids. <B>Each system requires a starter kit and clamp-on flow sensor; order separately.</B> The starter kit includes components to get a system in place, just choose a flow sensor at the desired flow range and sterility.
<b>Delivery information: </b>Includes water tight connection box.