Hubtron Transfer Tubing, Hubtron™ Black Pascal Fluoroelastomer, 1/8" ID x 1/4" OD; 25 Ft
Supplier: Avantor Fluid Handling
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
VWR Item#MFLX95257-02
Your Organization's Price:
Retrieving
Ideal for corrosive chemical transfer.
<B>Applications:</B> Extremely demanding corrosive chemical transfer, electronics applications, ink and printing, and general manufacturing applications requiring UV protection.<br /><B>Characteristics:</B> Opaque black with UV block, extremely low gas permeability, extremely strong chemical resistance. Superior pressure ratings. Excellent chemical resistance to solvents, acids, bases, alcohols, and esters.<br /><B>Certification:</B> None.<br /><B>Temperature:</B> 14 to 302 °F (–5 to 150 °C)<br /><B>Sterilization:</B> Not recommended.