Masterflex® Variable-Area Flowmeter, Direct-Read, Dual-Scale, 303 SS Housing, 10 GPM Water
Supplier: Avantor Fluid Handling
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
VWR Item#MFLX32205-14
Your Organization's Price:
Retrieving
Install in any position.
  • Direct-reading flowmeters feature a calibrated scale for a specific medium - no need to convert readings to get flow rate│Use with water for laboratory and industrial applications
Dual-scale flowmeters indicate gallons and liters simultaneously with an easy-to-read linear scale and highly visible red indicator.