Manual Pinch Valve, 7/16" to 1/2" tube OD
Supplier: Avantor Fluid Handling
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
An ideal option to secure tubing.
- Constructed of durable Delrin®
Fluid contacts only the tubing—ideal for use in pharmaceutical and food industries where eliminating contamination is critical. Obtain precise and repeatable flow rates using easy-to-read metered scale on valve body. Valves are made of Delrin®. Tubing is not included—order flexible tubing with durometer (Shore A) of 60 or less from our "Tubing" section.