Polypropylene flexible liner pump, rubber liner, 1 GPM, 115/230 VAC
Supplier: Avantor Fluid Handling
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
VWR Item#MFLX79440-00
Your Organization's Price:
Retrieving
Unique sealless design permits gentle pumping action that provides little churning or foaming.
  • Transfer liquids containing up to 20% soft solids
  • Sealless design with secondary containment system prevents potentially hazardous leakage
  • Fixed-speed pumps are self-priming, reversible, and can run dry without damage
Polypropylene pumps have a Nordel® rubber liner; PTFE pumps have a Hypalon® rubber liner. Variable-speed models and DC-powered pumps are available — call our Applications Specialists for more information.