Masterflex® Adhesive Coated Microscope Slide, Poly-l-lysine; 144/PK
Supplier: Avantor Fluid Handling
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
VWR Item#MFLX75955-45
Your Organization's Price:
Retrieving
Don't let your samples slip away.
  • Adhesive coatings fix cells and tissues to slides for hybridization and other staining techniques
  • Extensive quality control methods ensure each batch performs optimally
  • Coating formulations are effective without interfering with staining
Use these adhesive-coated microscope slides when sample adhesion is critical for proper staining and reliable results.<br /><br />A. Gelatin is the most common adhesive coating used in histology, ideal for simple stains.<br /><br />B. Poly-l-lysine is a stickier coating for more advanced immunostaining, but may result in precipitation during staining by silver impregnation methods.<br /><br />C. Silane offers superior adhesion, perfect for applications requiring antigen retrieval, and resists bleaching and enzymatic predigestion.<br /><br />D. Hybridization slides feature an adhesive coating that withstands organic solvents and a variety of temperatures, best suited to in situ hybridization applications.