Nylon manifold; 6 outlets; 1/4" inlet NPT(F)
Supplier: Avantor Fluid Handling
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
VWR Item#MFLX31522-43
Your Organization's Price:
Retrieving
Come in a variety of configurations—choose the number of ports and connections to suit your needs!
  • Constructed of durable nylon
Constructed of nylon for economic general applications while offering good chemial compatibility.