Flex Spout Adapter for 5-Gallon Containers with 2-5/8" Opening, polyethylene
Supplier: Avantor Fluid Handling
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
VWR Item#MFLX07079-15
Your Organization's Price:
Retrieving
Compatible with Siphon and Piston Hand-Operated Pumps.
  • Choose from a wide range of replacement parts and accessories
Model 07079-10 steel screw in adapter features a 1-3/4" NPT(F) connection and fits 5 gallon pails. <br />Model 07079-15 flex spout adapter is constructed of durable polyethylene and fits 5 gallon containers with 5-5/8" (6.7 cm) openings.<br />Model 07079-17 bung converter kit converts a Mauser course bung thread or standard course buttress to a 2" NPT connection.