Bev-A-Line Transfer Tubing, Bev-A-Line® IV, 1/8" ID x 1/4" OD; 50 Ft
Supplier: Avantor Fluid Handling
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
VWR Item#MFLX06490-12
Your Organization's Price:
Retrieving
Nontoxic and nonhemolytic.
  • Ideal for general-purpose, single-use, and extended fluid path applications
Applications: Safe for food, beverage, drug, lab, and general use—yet much more affordable than PTFE tubing. Safe for most alcohols; avoid use with strong solvents.<br />Characteristics: Polyethylene liner with ethyl vinyl acetate shell. No plasticizers. Nontoxic and nonhemolytic. Heat-bondable. Translucent. <br />Certification: Resin is FDA-compliant (liner: 21 CFR 177.1520; shell 21 CFR 177.1350). Meets USP Class VI, USDA, and National Formulary requirements.<br />Temperature range: –30 to 160 °F (–51 to 71 °C)<br />Sterilize: With ethylene oxide or dry heat.