Masterflex® Fitting, Polypropylene, Straight, Compression to Thread, 1/4" OD x 1/8" NPT(F); 10/PK
Supplier: Avantor Fluid Handling
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
VWR Item#MFLX06384-10
Your Organization's Price:
Retrieving
Unavailable
Maintain a permanent connection with these adapters.
  • Determine which connector type is needed for your application by contacting free technical application assistance*
  • Global market leader in range of materials for most extensive chemical compatibility
  • Single-use custom configuration solutions to minimise downtime
  • Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
  • Very good chemical resistance
  • Rated to 220 psi at 77°F
  • Temperature range of 0 to 275°F (-17 to 135°C)
  • Sterilized by ethylene oxide or autoclave
  • Connection Type: 1/4" OD x 1/8" NPT(F)
These threaded fittings are suitable for permanent installation and can used with your rigid tubing for high-pressure applications.