Masterflex® Fitting, Nylon, Straight, Compression to Thread Adapter, 1/4" OD x 1/8" NPT(F); 10/PK
Supplier: Avantor Fluid Handling
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
VWR Item#MFLX06382-57
Your Organization's Price:
Retrieving
Unavailable
Maintain a permanent connection with these adapters.
  • Determine which connector type is needed for your application by contacting free technical application assistance*
  • Global market leader in range of materials for most extensive chemical compatibility
  • Single-use custom configuration solutions to minimise downtime
  • Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
  • Good chemical resistance
  • Rated to 220 psi at 77°F
  • Temperature range of -40 to 200°F (-40 to 93°C)
  • Sterilized by ethylene oxide or autoclave
  • Connection Type: 1/4" OD x 1/8" NPT(F)
These threaded fittings are suitable for permanent installation and can used with your rigid tubing for high-pressure applications.