Masterflex® Microbore Transfer Tubing, Tygon® ND-100-80, 0.030" ID x 0.090" OD; 500 Ft
Supplier: Avantor Fluid Handling
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
VWR Item#MFLX06420-03
Your Organization's Price:
Retrieving
High resistance to kinking & collapse.
  • Ideal for general-purpose, single-use, and extended fluid path applications
  • Single-use custom lengths and configurations available to minimise downtime
  • Tubing size: 0.030" ID x 0.090" OD
<B>Formulation:</B> Tygon ND-100-80<br /><B>Applications:</B> General laboratory and pharmaceutical use, especially with precision injection or dispensing applications. Also excellent with low-flow peristaltic pump applications.<br /><B>Characteristics:</B> Extremely flexible. Non-toxic; nonpyrogenic; biocompatible. Crystal clear.<br /><B>Certifications:</B> Resin meets USP Class VI requirements, non-DEHP.<br /><B>Temperature range:</B> -31 to 185 °F (-35 to 85 °C)<br /><B>Sterilize:</B> Autoclave, ethylene oxide, or gamma irradiation.