Masterflex® B/T® Analog Air-Powered Drive, 35-321 rpm; 30 CFM at 60 PSI
Supplier: Avantor Fluid Handling
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
VWR Item#MFLX77111-80
Your Organization's Price:
Retrieving
High-volume transfer and dispensing.
  • Flow rate 0.71/2.1 to 42 LPM with B/T tubing—flow rate depends on drive rpm and tubing size
  • Rapid-Load® pump head occlusion mechanism slides occlusion bed out of the way for easy tube loading and improved tubing occlusion
  • Accepts B/T PerfectPosition™ pump tubing sizes B/T 87 and B/T 91
  • Interlock switch shuts off power to stop drive when door is opened
  • Use air-powered drive in locations where electricity is unsafe or impractical
  • Includes pressure regulator — control your compressor air to match pump requirements
  • 24 cfm (0.68 m³), 100 psi (6.9 bar) minimum air required to achieve max rated flow
  • Rated for ATEX Zone 2, EEx II 3 G c IIC T6; NEC rated to Class 1, Division 2, Groups A, B, C, and D, T6
  • IP56-rated motor protects against dust and strong jets of water
  • Compressed air input requirement is 30 cfm at 60 psig
  • Use air-powered drive in locations where electricity is unsafe or impractical
  • Includes pressure regulator — control your compressor air to match pump requirements
  • Rated for ATEX Zone 2, EEx II 3 G c IIC T6; NEC rated to Class 1, Division 2, Groups A, B, C, and D, T6
Masterflex® B/T® drives are the best choice for high-flow bulk-transfer applications. Their durable construction ensures long service life with minimal maintenance. The sturdy materials of construction include stainless steel/carbon steel, a cast aluminum housing with an epoxy-powder coating, and Nylatron® rollers for rugged dependability.<br /><br />All B/T drives include a new and improved Rapid-Load® pump head for fast and easy tubing changes. The redesigned occlusion mechanism provides an improved tubing retention mechanism which holds the tubing firmly in place to further simplify the loading process. PerfectPosition™ tubing sizes provide improved performance in higher-pressure applications and provide longer tubing life.
<b>Delivery information: </b>Includes rapid-Load pump head and 3 ft (1 m) of B/T 91 tubing. Order additional tubing under the Masterflex tubing section, B/T PerfectPosition Tubing.
<b>Delivery information: </b>Includes rapid-load pump head and 3 ft (1 m) of B/T 91 tubing.