Masterflex® Fitting, HDPE, Straight, Hose Barb Union, 1/16" ID;10/PK
Supplier: Avantor Fluid Handling
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
VWR Item#MFLX40612-01
Your Organization's Price:
Retrieving
Unavailable
Quick and easy assembly without tools.
  • Determine which connector type is needed for your application by contacting free technical application assistance*
  • Global market leader in range of materials for most extensive chemical compatibility
  • Single-use custom configuration solutions to minimise downtime
  • Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
  • Chemical resistant
  • Connection Type: 1/16" ID with L/S 13/14