Masterflex® Ismatec® Minicartridge Multichannel Pump Head for Masterflex® L/S® Drives, 8-Channel, 8-Roller
Supplier: Avantor Fluid Handling
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
VWR Item#MFLX07623-10
Your Organization's Price:
Retrieving
Combine the accuracy of Ismatec® with the versatility of Masterflex® tubing pump systems.
  • Order optional minicartridges with adjustable occlusion to vary pressure each channel
  • Easily remove or insert minicartridges without disturbing other channels
  • Max pressure of 1.5 bar is achieved with pressure-lever cassettes only
  • Click-n-go cassettes offer a max pressure of 1.0 bar.
  • Maximum flow rate at 100 rpm is 28.5 mL/min
  • 8-channel pump head features 8 rollers
Choose from six, eight, or twelve roller heads. Less rollers offer greater flow rates, while more rollers offer a virtually pulseless flow. Heads accept up to four or eight Click’n’go™ minicartridges (included). Click’n’go minicartridges feature spring occlusion for optimum tension at all times with no adjustments.<br /><br />Call our Application Specialists to determine the maximum recommended number of cartridges to use with your drive.
<b>Delivery information: </b>Includes eight Click’n’go™ minicartridges.