Masterflex® Footswitch, Momentary Start/Stop Operation, DB25; 6 ft
Supplier: Avantor Fluid Handling
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
VWR Item#MFLX07523-92
Your Organization's Price:
Retrieving
Easily control your Masterflex® drive with compatible footswitch.
  • Choose footswitch with the correct connection for your Masterflex® pump
  • Connection Type: DB25
Add convenience to repetitive dispensing with a foot-operated on/off switch. Keep hands free to efficiently move containers during routine fill applications or to keep containers stable during the filling process.