Masterflex® Ismatec® Pump Tubing, 2-Stop, Platinum-Cured Silicone, 0.89 mm ID; 6/PK
Supplier: Avantor Fluid Handling
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
VWR Item#MFLX06421-26
Your Organization's Price:
Retrieving
Excellent biocompatibility that meets multiple classifications.
  • Genuine Ismatec® tubing undergoes robust testing to provide optimal performance / pump life hours with peristaltic pump equipment
  • Precision extruded to tight tolerances for high repeatability, accuracy and reproducibility
  • Single-use custom lengths and configurations available to minimise downtime
  • Tubing Inner Diameter: 0.89 mm
Compatible with Masterflex® L/S® Multichannel pump head, small cartridges.<br /><br />Applications: Pharmaceutical, biotech, peristaltic pumping, and food applications<br /><br />Characteristics: Excellent heat resistance, low compression set, odorless, nontoxic, translucent<br /><br />For custom tubing lengths and configurations, please contact our Product Applications Specialist.