Masterflex® I/P® Spooled Precision Pump Tubing, C-Flex®, I/P 82; 100 ft
Supplier: Avantor Fluid Handling
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
VWR Item#MFLX06427-82
Your Organization's Price:
Retrieving
Maintain flexibility at extreme temperatures.
  • Genuine Masterflex® tubing undergoes robust testing to provide optimal performance/pump life hours with peristaltic pump equipment
  • Precision extruded to tight tolerances for high repeatability, accuracy, and reproducibility
  • Single-use custom lengths and configurations available to minimise downtime
  • Ultra smooth inner surface results in fewer extractables
  • I/P® 82 tubing has a 1/2” (12.7 mm) inner diameter
  • Accepts 1/2” hose barb for tubing connection
Masterflex® C-Flex® Precision pump tubing offers enhances biocompatibility with longer pumping life. This tubing is nonpyrogenic, noncytotoxic, and nonhemolytic — protecting the purity of your fluid. Heat sealable and weldable, C-Flex tubing is ideal for biopharmaceutical processing and storage and botanical production.<br /><br />Spools contain continuous pump tubing, allowing you to cut the exact length needed and eliminating waste.