Masterflex® L/S® High-Performance Precision Pump Tubing, FDA-Compliant Viton®, L/S 24; 25 ft
Supplier: Avantor Fluid Handling
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
VWR Item#MFLX96412-24
Your Organization's Price:
Retrieving
Good for food and lab applications.
  • Genuine Masterflex® tubing undergoes robust testing to provide optimal performance / pump life hours with peristaltic pump equipment
  • Precision extruded to tight tolerances for high repeatability, accuracy and reproducibility
  • Single-use custom lengths and configurations available to minimise downtime
  • Unique packaging options include packs, cases, and spools for customer-friendly ordering and delivery
  • Tubing Size: L/S 24
<B>Applications</B>: Food or laboratory applications where FDA compliance is required, acid and solvent transfer, high-temperature work<br /><B>Characteristics</B>: Good chemical resistance, opaque black