Container, EUROBOX
Supplier: ZARGES LEICHTBAU
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
40700 40706 40702 40709
ZARG40700EA 326 GBP
ZARG40700 ZARG40706 ZARG40702 ZARG40709
Container, EUROBOX
Boxes Storage Boxes
Aluminium, with lid.
  • Lid with stainless steel hinges and holding straps
  • Blue stacking corners ensure safe stacking
  • Corner beads for additional dimensional stability
Eurobox has high resistance against corrosion due to stainless steel fittings. The peripheral foamed seal in the lid protects against dust and splash water.
New Product
Order Now
SPECIFICATIONS
Environmentally Preferable