Conductivity standard 10,000 µS/cm (25°C; 0,0765 mol/l; KCl) for conductivity measurement
Supplier: YSI ENVIRONMENTAL
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
060911
YSIA060911EA 54.7 GBP
YSIA060911
Conductivity standard 10,000 µS/cm (25°C; 0,0765 mol/l; KCl) for conductivity measurement
Standards Conductivity Standards