Trypsin
Supplier: BIOWEST
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
L0910-100 L0930-100 L0930-500 L0940-100 L0940-500 P0940-1GR P0940-100GR X0915-100 X0930-100 P5957-100GR L0909-100 L0931-100 L0931-500 L0932-100 X0920-100 L0941-100
L0910-100EA 14.8 GBP
L0910-100 L0930-100 L0930-500 L0940-100 L0940-500 P0940-1GR P0940-100GR X0915-100 X0930-100 P5957-100GR L0909-100 L0931-100 L0931-500 L0932-100 X0920-100 L0941-100
Trypsin
Cell Culture Supplements and Additives
Trypsin is a porcine pancreas-derived enzyme that is commonly used for the dissociation and disaggregation of anchorage-dependent mammalian cells and tissues. The concentration of trypsin necessary to dislodge the cells from their substrate depends on the sensitivity of the cells.
Order Now
SPECIFICATIONS
Environmentally Preferable
Species
Source
Learn more